Bone-Cutting Shears
Bone-Cutting Shears
Actual product may vary
Fisherbrand
Bone-Cutting Shears
Heavy-duty shears that cut through bones.
Have Questions?
Catalog NumberLength (English)
FISS173425.5 in.
Catalog number FISS17342
Price (SGD)
-
Length (English):
5.5 in.
  • Made of stainless steel, the small shear is ideal for small bones
  • The large shears cut large bones easily and cleanly
Specifications
Blade StyleStraight
Length (English)5.5 in.
Length (Metric)14 cm
For Use With (Application)Surgical, Bone
Unit SizeEA