▸
Blood, Hematology and Coagulation Testing Products
▸
Cell Counting Devices
▸
Hemocytometer
Actual product may vary
Hemocytometer
Get quick, accurate readings of red and white blood cells.
Have Questions?
Catalog Number
For Use With (Application)
FISS17040
Tool for Blood tests
Catalog number
FISS17040
Price (SGD)
-
For Use With (Application):
Tool for Blood tests
Easy-to-read, brightline Neubauer counting chamber makes device an important tool for blood tests
Made from one piece of optical glass, its double ruling allows check counts to be made for accuracy
Specifications
Product Type
Hemacytometer
Grid Type
Neubauer Bright Line
Quantity
Each
For Use With (Application)
Tool for Blood tests
Includes
Two cover Glass, one red pipette, one white pipette, one pipette filler