Test Tube Brush
Test Tube Brush
Actual product may vary
Test Tube Brush
Test Tube Brush with fan-shaped end.
Have Questions?
Catalog NumberLength (Metric)DiameterMetric
FISS1395823.49 cm
Catalog number FISS13958
Price (SGD)
-
Length (Metric):
23.49 cm
  • On stainless-steel wire handle
Specifications
Product TypeBristle Brush
Bristle MaterialFiber
Bristle TypeMedium Stiffness
Color-codedYes
Handle MaterialStainless Steel Wire
Handle TypeTwisted Wire
Tip TypeFan Tip
ScentUnscented
SterilityNon-sterile
ColorTan
Diameter (English) Bristle0.5 in.
Diameter (Metric) Bristle12.7 mm
Height (English)0.5 in.
Height (Metric)1.27 cm
Length (English)9.25 in.
Length (English) Brush3.8 in.
Length (Metric)23.49 cm
Length (Metric) Brush95.2 mm
Quantity1 Ea.
Weight (English)0.02 lb.
Weight (Metric)9 g
Width (English)0.5 in.
Width (Metric)1.27 cm
Unit SizeEA