Check Order Status
Small Graduated Cup - 25mL capacity, 5mL graduations
Actual product may vary
Small Graduated Cup - 25mL capacity, 5mL graduations
These transparent polypropylene cups can be used to measure different volumes up to 25mL.
Have Questions?
Catalog Number
Capacity (Metric)
FISS13947
25 mL
Catalog number
FISS13947
Price (SGD)
-
Capacity (Metric):
25 mL
Specifications
Quantity
250/Pk.
Capacity (Metric)
25 mL
Product Type
Sample Cup
Capacity (English)
0.84 oz.
Material
Polyethylene
Capacity
5 mL