Premium Large Three Finger Clamp w/ Boss head
Premium Large Three Finger Clamp w/ Boss head
Actual product may vary
Premium Large Three Finger Clamp w/ Boss head
Choose durable lab equipment that will last for years. The EISCO Premium Large Three Finger Clamp w/ Boss head is an industrial clamp that has rubber coated fingers.
Have Questions?
Catalog NumberColor
FISS13914Blue
Catalog number FISS13914
Price (SGD)
-
Color:
Blue
  • Clamp constructed of powder coated steel, 3 finger rubber coated clamp attached to aluminum rod
  • Maximum clamp opening is 8.5 cm
  • 14 cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10 mm diameter connecting rod
Specifications
AutoclavableAutoclavable
Type3-Prong Extension Clamp
MaterialZinc Die Cast, Powder Coated
ColorBlue
Jaw Type3-Prong
CoatingRubber
Unit SizeEA