Premium Two Prong Clamp
Actual product may vary
Premium Two Prong Clamp
Choose durable lab equipment that will last for years. The Premium Two Prong Clamp is an industrial two prong clamp for burettes, flasks, and beaker clamps.
Have Questions?
Catalog NumberColor
FISS13909Blue
Catalog number FISS13909
Price (SGD)
-
Color:
Blue
  • Clamp constructed of powder coated zinc alloy, 2 prongs attached to aluminum rod
  • Maximum clamp opening is 12 cm
  • 14 cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10 mm diameter connecting rod
Specifications
AutoclavableAutoclavable
Type2-Prong Clamp
MaterialNickel-plated
ColorBlue
Jaw Type2-Prong
CoatingVinyl
Unit SizeEach