Premium Four Prong Clamp with Boss Head
Actual product may vary
Premium Four Prong Clamp with Boss Head
Choose durable lab equipment that will last for years. The Premium Four Prong Clamp with Boss Head is an industrial, rubber-lined universal lab clamp with swivel boss head attachment.
Have Questions?
Catalog NumberColor
FISS13906Blue
Catalog number FISS13906
Price (SGD)
-
Color:
Blue
  • Clamp constructed of powder coated steel
  • Maximum clamp opening is 10.5 cm
  • Swivel between clamp and boss head
  • Boss accommodates up to 19 mm rod
Specifications
AutoclavableAutoclavable
Quantity1
Connector TypesConnecting Rod
TypeAdjustable Clamp
MaterialZinc Die Cast, Powder Coated
IncludesBoss Head
For Use With (Equipment)Beakers, Erlenmeyer flasks, test-tubes, boiling (Florence) flasks, burettes
DescriptionVinyl Coated
Height (Metric)25.4 mm
Height (English)1 in.
Length (English)8 in.
Length (Metric)203.2 mm
Width (English)4 in.
Width (Metric)101.6 mm
ColorBlue
Jaw Type4-Prong
SterilityNon-sterile
CoatingVinyl
Unit SizeEach