Premium Four Prong Clamp
Premium Four Prong Clamp
Actual product may vary
Fisherbrand
Premium Four Prong Clamp
Choose durable lab equipment that will last for years. The EISCO Premium Four Prong Clamp is an industrial universal lab clamp.
Have Questions?
Catalog NumberColor
FISS13903Blue
Catalog number FISS13903
Price (SGD)
-
Color:
Blue
  • Clamp constructed of powder coated steel
  • Lined clamp prongs are attached to aluminum rod
  • Maximum clamp opening is 10.5 cm
  • 14cm connecting rod
  • Completely metal constructions, thread fitted actuating joints
  • 10mm diameter connecting rod
Specifications
AutoclavableNot Autoclavable
TypeAdjustable Clamp
MaterialZinc Die Cast, Plastic Coated
Height (Metric)2.54 cm
Height (English)1 in.
Length (English)9.5 in.
Length (Metric)24.13 cm
Width (English)4 in.
Width (Metric)10.16 cm
ColorBlue
Jaw Type4-Prong
CoatingCork
Unit SizeEA