Cobalt Glass Plates
Actual product may vary
United Scientific Supplies
Cobalt Glass Plates
Cobalt Glass Plates used in flame tests to block orange color when performing sodium and potassium salt flame tests.
Have Questions?
Catalog NumberDimensions (L x W)
FISS1313810.1 x 10.1 cm (4 x 4 in.)
Catalog number FISS13138
Price (SGD)
-
Dimensions (L x W):
10.1 x 10.1 cm (4 x 4 in.)
  • Color: Dark blue
  • Available in three different sizes
Specifications
ColorDark Blue
For Use With (Equipment)Bunsen Burner
MaterialCobalt Glass
SterilityNon-sterile
TypeCobalt Plate
Dimensions (L x W)10.1 x 10.1 cm (4 x 4 in.)
Length (English)4 in.
Length (Metric)10.1 cm
Width (English)4 in.
Width (Metric)10.1 cm
Unit SizePack