Stainless Steel Laboratory Scoop
Stainless Steel Laboratory Scoop
Actual product may vary
Fisher Catalogue
Stainless Steel Laboratory Scoop
Stainless Steel Laboratory Scoop designed for transferring powdered or granular materials.
Have Questions?
Catalog NumberCapacity (English)
FISS1308452 oz.
Catalog number FISS13084
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (English):
52 oz.
  • For general laboratory use.
  • Available in two sizes: 5 and 52 oz.
Specifications
Quantity1 Ea.
AutoclavableAutoclavable
ShapeScoop
Capacity (English)52 oz.
Capacity (Metric)1474 g
Product TypeLaboratory Scoop
MaterialStainless Steel
Length (Metric)419 mm
Unit SizeEA