Secchi Disk Including Rope
Secchi Disk Including Rope
Actual product may vary
Secchi Disk Including Rope
The Secchi Disk Including Rope is used to determine transparency of water in lakes, streams, oceans.
Have Questions?
Catalog NumberGrade
FISS130199 to College
Catalog number FISS13019
Price (SGD)
-
Grade:
9 to College
  • Has nine in. disc of alternating black and white quadrants
  • Provides quick and easy way to probe turbidity, which is measure of clarity of water
  • Turbidity is in turn related to amount of sediments or pollutants in water
Specifications
Description9 in. disc with black and white quadrants
IncludesSecchi Disk, 10+ Ft. of String, Small Handle
Grade9 to College
Product TypeSecchi Disk
For Use With (Application)Quick qualitative measurements of the clarity of water over time
Unit SizeEA