Wire Gauzes with Ceramic Center
Actual product may vary
United Scientific Supplies
Wire Gauzes with Ceramic Center
Used for general heating purposes in laboratories.
Have Questions?
Catalog NumberDimensions (L x W)
FISS1286612.7 x 12.7 cm (5 x 5 in.)
Catalog number FISS12866
Price (SGD)
-
Dimensions (L x W):
12.7 x 12.7 cm (5 x 5 in.)
  • Iron wire gauze (24 SWG/10 Mesh) with folded edges
  • Contains no asbestos
  • Sold in packs of 10
  • Available in two different sizes
Specifications
Dimensions (L x W)12.7 x 12.7 cm (5 x 5 in.)
EdgesFolded
IncludesCeramic Center
Length (English)5 in.
Quantity10/Pk.
ShapeSquare
Width (English)5 in.
Length (Metric)12.7 cm
MaterialIron
Width (Metric)12.7 cm
Product TypeWire Gauze
Unit SizePack