Check Order Status
Neoprene Hose for Bunsen Burner
Actual product may vary
Neoprene Hose for Bunsen Burner
High quality neoprene tubing to connect Bunsen burners to fuel source.
Have Questions?
Catalog Number
Length (English)
Length (Metric)
FISS12817
2 ft.
0.6 m
Catalog number
FISS12817
Price (SGD)
-
Length (English):
2 ft.
Length (Metric):
0.6 m
Connect your Bunsen burners to a fuel source such as natural gas or liquid propane
Both ends of the hose are designed to fit gas inlet diameters of roughly 8 to 10 mm
Specifications
Product Type
Gas Hose
For Use With (Equipment)
Bunsen burner
Gas Type
Natural Gas/Propane
Length (English)
2 ft.
Length (Metric)
0.6 m
Material
Neoprene
Color
Blue
Diameter (Metric) Inside
8 to 10 mm
For Use With (Application)
Connect burners to fuel source