▸
Racks
▸
Tube Racks
▸
Six Hole Wood Test tube Rack
Actual product may vary
Six Hole Wood Test tube Rack
Six Hole Wood Test tube Rack with coated finish to protect from chemicals.
Have Questions?
Catalog Number
Positions
FISS07854
6
Catalog number
FISS07854
Price (SGD)
-
Positions:
6
Made of wood.
Holds six tubes with 20 mm holes.
Specifications
Autoclavable
Autoclavable
Color
Brown
Holds
Tubes (24 mm)
Material
Wood
Positions
6
Shape
Vertical
Stackable
Yes, When Empty
Sterility
Non-sterile
Diameter (Metric) Opening
24 mm
Product Type
Tube Rack
Shelves
2 Tiers
Array
6 holes