Plastic Well Slides
Actual product may vary
United Scientific Supplies
Plastic Well Slides
These Plastic Well Slides are a reusable alternative to the glass slide with well.
Have Questions?
Catalog NumberNo. per Pack
FISS0736510 Pk.
Catalog number FISS07365
Price (SGD)
-
No. per Pack:
10 Pk.
  • Reusable.
  • Dimension: 7.6 x 2.5 cm (3 x 1 in.)
  • Well dimensions: approximately 16 mm diameter with a depth of 2 to 3 mm.
  • Construction: clear plastic
  • Supplied as sheets of 10.
Specifications
No. per Pack10 Pk.
FrostedNot Frosted
Length (Metric)7.6 cm
MaterialPlastic
PrecleanedNo
Thickness (Metric)2 to 3 mm
TypeWell Slide
Well Count1
Width (Metric)2.5 cm
ColorClear
DescriptionWell is approximately 16 mm diameter with a depth of 2 to 3 mm
Length (English)3 in.
Width (English)1 in.
Dimensions (L x W)7.6 x 2.5 cm (3 x 1 in.)
Unit SizePack