Botany Dissecting Kit in a Zippered Case
Botany Dissecting Kit in a Zippered Case
Actual product may vary
Fisherbrand
Botany Dissecting Kit in a Zippered Case
This Botany Dissecting Kit is an ideal choice for field work and lab use.
Have Questions?
Catalog number FISS06839
Price (SGD)
-
  • Dissecting kit for botany students, teachers and field botanists
  • Superior quality dissection tools in zippered vinyl case
  • Contains all the necessary tools required for plant study, dissection and analysis
  • All tools are made with stainless steel unless otherwise noted
Includes:
  • Dissecting iris scissors, 4.5 in.
  • Two dissecting forceps, 4.5 in. with guide pin (one curved fine point, one medium point)
  • Two teasing needles (one angular, one straight) with metal chuck, brass-chromed
  • Dissecting scalpel handle No. 3
  • No. 10 blade (10 pcs.) or No. 15 blade
  • Ruler, 6 in.
  • Glass dropper
  • Replacement dissecting needles, straight and angular (pack of six)
  • Vinyl zippered case with linen (black or red)
Specifications
Case MaterialVinyl
For Use With (Application)An excellent choice for field work and lab use
IncludesDissecting Iris Scissors, 2 Dissecting Forceps, 2 Teasing Needles, Dissecting Scalpel Handle No. 3, No. 10 Or No. 15 Blade, Ruler, Glass Dropper, 6 Replacement Dissecting Needles, Vinyl Zippered Case With Linen
MaterialStainless Steel
Quantity1 Ea.
Product TypeBotany Dissecting Kit
Unit SizeEA