Metalware Set
Actual product may vary
Metalware Set
Metalware Set
Metalware Set
One each base, rod, boss head, clamp and retort ring.
Have Questions?
Catalog NumberHeight (Metric)
FISS026311 in.
Catalog number FISS02631
Price (SGD)
-
Height (Metric):
1 in.
One each base, rod, boss head, clamp and retort ring.
  • The set includes:
    • Base retort stand 20 x 12.5 cm (Fisher Cat S02611)
    • Steel rods, 60 cm (Fisher Cat S02615)
    • Boss head (Fisher Cat S24263)
    • 4-prong powder coated clamp retort (Fisher Cat S24275)
    • Retort ring, 75 mm O.D.
Specifications
MaterialSteel (Rod)
Height (Metric)1 in.
IncludesOne each base 20 x 12.5 cm, 60 cm rod, boss head, clamp and retort ring
Product TypeMetalware Set
ShapeRectangular Base
QuantityEach
Unit SizeEach