Stainless Steel Rods for Retort Base Stands
Actual product may vary
Stainless Steel Rods for Retort Base Stands
Stainless steel rods for retort base stands have threaded bottoms and will resist corrosion and stains from chemicals.
Have Questions?
Catalog NumberLength (Metric)
FISS0262475 cm
Catalog number FISS02624
Price (SGD)
-
Length (Metric):
75 cm
  • Sturdy stainless steel rods
  • Include threaded end for use with retort stands
  • Corrosion and chemical resistant
  • Reliable in a lab setting when used with a base
Specifications
MaterialStainless Steel
Height (Metric)2.54 cm
Height (English)1 in.
Length (English)29.5 in.
Length (Metric)75 cm
Width (English)5.3 in.
Width (Metric)13.46 cm
Diameter (English) Inner0.47 in.
Diameter (English) Rod0.47 in.
Diameter (Metric) Rod12 mm
Product TypeRod
Thread Size10 x 1.5 mm
For Use With (Equipment)Base retort stand
Weight (English)1.5 lb.
Weight (Metric)680.38 g
Unit SizeEach