▸
Clamps and Supports
▸
Stands and Rings
▸
Base Retort Stand
Actual product may vary
Base Retort Stand
Rectangular in shape.
Have Questions?
Catalog Number
Height (English)
Length (English) Base
Weight (English)
Width (English) Base
FISS02610
1.5 in.
6.2 in.
2.4 lb.
3.9 in.
Catalog number
FISS02610
Price (SGD)
-
Height (English):
1.5 in.
Length (English) Base:
6.2 in.
Weight (English):
2.4 lb.
Width (English) Base:
3.9 in.
Pressed steel base
With hole 10 x1.5mm metric thread in the center of one short side
Finished in blue/grey colour
Specifications
Material
Steel
Height (Metric)
4 cm
Height (English)
1.5 in.
Length (English) Base
6.2 in.
Length (Metric) Base
16 cm
Width (English) Base
3.9 in.
Width (Metric) Base
10 cm
Product Type
Base Retort Stand
Shape
Rectangular
Thread Size
0.39 x 0.05 in.
Color
Blue/Gray
Quantity
Each
Weight (English)
2.4 lb.
Weight (Metric)
1 kg