Azlon™ Polypropylene Carboy with Spigot
Actual product may vary
Dynalon
Azlon™ Polypropylene Carboy with Spigot
For Use With: Storing and dispensing liquids in educational settings.; Autoclavable; Closure Material: Polypropylene; Color: White
Have Questions?
Catalog NumberCapacity (Metric)
FISS019895 L
Catalog number FISS01989
Price (SGD)
-
Capacity (Metric):
5 L
  • Autoclavable polypropylene™, heavy-duty construction
  • Stylish rounded octagonal shape for increased rigidity when full
  • Offset neck for easier filling and pouring
  • Integral, comfortable blow molded handles for ease of carrying, are end-sealed to prevent contamination
  • Heavy-duty closures
  • Carboys feature a durable and leakproof heavy-duty spigot with boss fitting for extra strength
  • Spigot features removable tube fitting for added convenience
  • Molded graduations allow easy viewing of contents
Specifications
AutoclavableAutoclavable
Capacity (English)1.3 gal.
Capacity (Metric)5 L
Closure MaterialPolypropylene
ColorWhite
For Use With (Application)Storing and dispensing liquids in educational settings.
IncludesBlow molded handles
MaterialPolypropylene
Neck StyleOffset
Product TypeCarboy with Spigot
ShapeRounded Octagonal
Unit SizeEach