Kartell™ Cylindrical Jars
Kartell™ Cylindrical Jars
Actual product may vary
Fisherbrand
Kartell™ Cylindrical Jars
Suitable for powders and solid specimens.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FISS01987D1000 mL110 mm
Catalog number FISS01987D
Price (SGD)
-
Capacity (Metric):
1000 mL
Closure Size:
110 mm
  • HDPE jars with large necks
  • Supplied with insert and HDPE cap
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
Diameter (English)4.4 in.
For Use With (Application)Powders and Solid Specimens
IncludesScrew Cap
MaterialHDPE
AutoclavableNot Autoclavable
ShapeRound
Height (Metric)128 mm
Product TypeJar
Height (English)5 in.
Closure MaterialPolypropylene
Closure Size110 mm
Diameter (Metric)111 mm
Closure TypeScrew Cap
Unit SizePK