1x1 image pixel for data collection
Visitorspec Safety Glasses
Visitorspec Safety Glasses
Actual product may vary
Visitorspec Safety Glasses
Ideal for protecting visitors in industrial or medical settings.
Have Questions?
Catalog NumberLens Coating
FISS01819Anti-fog, Anti-static, Scratch-resistant
Catalog number FISS01819
Price (SGD)
-
Lens Coating:
Anti-fog, Anti-static, Scratch-resistant
Economical safety spectacles available with coated or uncoated lens.
Uncoated Lens
  • 100% impact-resistant polycarbonate wraparound unilens design with blue temples
  • Clear lens absorbs >99.9% of harmful UV light
Coated Lens
  • 100% impact-resistant lens absorbs >99.9% of harmful UV light
  • Polycarbonate wraparound style with molded-in browguard and single lens
  • Coated to resist scratching, fogging and static
  • Adjustable-length spatula temples ensure a secure, comfortable fit
Specifications
Lens CoatingAnti-fog, Anti-static, Scratch-resistant
Certifications/ComplianceMeet ANSI Z87.1-2003 standards.
Frame ColorBlue
No. per Pack12/Pk.
Temple StyleAdjustable-length; spatula
Lens MaterialPolycarbonate
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link