Check Order Status
Starch Indicator, 0.5% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
Element Search
Structure Search
Starch Indicator, 0.5% (w/v), Mercury Free, for Iodometric Titrations, Ricca Chemical
Iodometric Titrations
Have Questions?
Change view
Quantity:
100 mL
1 L
Packaging:
Natural Poly Bottle
Catalog number
FISR8000000100
Price (SGD)
-
Quantity:
100 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
9005-84-9, 69-72-7, 7732-18-5
Specifications
Chemical Name or Material
Starch Indicator
CAS
9005-84-9
CAS
69-72-7
CAS
7732-18-5
Concentration
0.5% (w/v)
View more