Check Order Status
Hydroxylamine Hydrochloride Solution, USP <251> for Lead, Ricca Chemical
Element Search
Structure Search
Hydroxylamine Hydrochloride Solution, USP <251> for Lead, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FISR39124
120 mL
Natural Poly Bottle
Catalog number
FISR39124
Price (SGD)
-
Quantity:
120 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Hydroxylamine hydrochloride solution
CAS
7732-18-5
CAS
67-66-3
CAS
5470-11-1
Physical Form
Liquid
View more