Check Order Status
DPD Indicator Solution, for Residual Chlorine Analysis, Ricca Chemical
Element Search
Structure Search
DPD Indicator Solution, for Residual Chlorine Analysis, Ricca Chemical
For Analysis of Residual Chlorine Analysis
Have Questions?
Catalog Number
Quantity
FISR2655000100
100 mL
Catalog number
FISR2655000100
Price (SGD)
-
Quantity:
100 mL
Request bulk or custom format
Chemical Identifiers
CAS
7732-18-5, 7664-93-9, 6283-63-2, 6381-92-6
Specifications
Chemical Name or Material
DPD Indicator Solution
CAS
7732-18-5
CAS Min %
98.0
CAS Max %
100.0
CAS
7664-93-9
View more