Check Order Status
Chloride Standard, 5 ppm Cl
-
, Ricca Chemical
Element Search
Structure Search
Chloride Standard, 5 ppm Cl
-
, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FISR19415001A
1 L
Natural Poly Bottle
Catalog number
FISR19415001A
Price (SGD)
0.00
EA
Add to cart
Quantity:
1 L
Packaging:
Natural Poly Bottle
Request bulk or custom format
Price (SGD)
0.00
EA
Add to cart
Specifications
Chemical Name or Material
Chloride Standard
Name Note
1mL = 0.005mg Cl-, 5ppm Cl-
Packaging
Natural Poly Bottle
CAS
7647-14-5
CAS
7732-18-5
View more