Check Order Status
Buffer, Reference Standard, pH 1.00 ± 0.01 at 25°C, Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 1.00 ± 0.01 at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FISR148916
500 mL
Natural Poly Bottle
Catalog number
FISR148916
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7647-01-0, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Name Note
pH 1.00± 0.01 at 25°C
CAS
7647-01-0
Color
Colorless
CAS
7732-18-5
View more