Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Filters and Filtration
▸
Filtration Membrane Discs
▸
Magna™ Nylon Membrane Filters
Actual product may vary
GVS
Magna™ Nylon Membrane Filters
Plain white nylon membranes offer low extractables, high uniformity
Have Questions?
Change view
Catalog Number
Pore Size
Diameter (Metric)
Surface Type
FISR02SP04700
0.2 μm
47 mm
Plain
FISR12SP04700
1.2 μm
47 mm
Plain
2 Options
Catalog number
FISR02SP04700
Price (SGD)
400.00
Pack
Add to cart
Pore Size:
0.2 μm
Diameter (Metric):
47 mm
Surface Type:
Plain
Price (SGD)
400.00
Pack
Add to cart
Plain white nylon membranes offer low extractables, high uniformity
Won't crack, tear, or distort in handling or creasing
Resist heat, sterilization, and most organic solvents
Hydrophilic, no wetting agents; extractable level <0.0015mg/cm
Heatable, autoclavable up to 180°C (356°F)
Broad chemical compatibility
Not compatible with strong acids or bases
Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC
For microbiological work, use 47mm/0.45μm size with grid
Specifications
Autoclavable
Autoclavable
Bubble Point
46psi
Material
Nylon
Pore Size
0.2 μm
Flow Rate
13.5mL/min./cm sq.
For Use With (Application)
Permit use for aqueous filtration without wetting and organic solvent filtrations such as HPLC
Gas Flow Rate
1.7L/min./cm sq.
Hydrophobicity
Hydrophilic
Membrane Type
Plain
Surface Type
Plain
Diameter (Metric)
47 mm