Shaker Flasks
Actual product may vary
Shaker Flasks
Reduce the risk of cross-contamination.
Have Questions?
Catalog NumberCapacity (Metric)Bottom ShapeClosure Type
FISPBV12-5125 mLPlainVented
Catalog number FISPBV12-5
Price (SGD)
441.00
Case
Capacity (Metric):
125 mL
Bottom Shape:
Plain
Closure Type:
Vented
Price (SGD)
441.00
Case
Reduce the risk of cross-contamination.
  • Nonpyrogenic and noncytotoxic
  • Clear, durable polycarbonate with molded-in graduations
  • Polypropylene closure with 0.22μm PTFE pore
  • Individually packed for easy storage and handling
  • Plain bottom for use on benchtop or baffled bottom for shaker table use
  • Vented or non-vented screw closure NOT autoclavable
  • Autoclavable flask
  • Sterile
Specifications
Capacity (English)4.22 oz.
Closure MaterialPolypropylene
ColorClear
Capacity (Metric)125 mL
MaterialPolycarbonate
For Use With (Application)For suspension cell culture, media preparation and storage
Includes0.22μm PTFE pore
AutoclavableAutoclavable
GraduatedYes
SterilitySterile
ClassClass VI
Bottom ShapePlain
Closure TypeVented
Unit SizeCase