▸
Salts and Inorganics
▸
Inorganic Salts
▸
Alkali Metal Salts
▸
Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Potassium chloride for calomel cells
Have Questions?
Catalog Number
Quantity
Packaging
FISP217-500
500 g
Poly Bottle
Catalog number
FISP217-500
Price (SGD)
-
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
1420°C
Chemical Name or Material
Potassium Chloride
Melting Point
770°C
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
View more