Oxoid™ Gelatin Bacteriological
Oxoid™ Gelatin Bacteriological
Actual product may vary
Fisherbrand
Oxoid™ Gelatin Bacteriological
Oxoid Gelatin Bacteriological is a special bacteriological grade readily soluble in water to give a clear solution free from sulphite and preservatives.
Have Questions?
Catalog NumberDescription
FISOXLP0008BDehydrated; Gelatin bacteriological
Catalog number FISOXLP0008B
Price (SGD)
-
Description:
Dehydrated; Gelatin bacteriological
Oxoid Gelatin Bacteriological is a special bacteriological grade readily soluble in water to give a clear solution free from sulphite and preservatives.
  • Firm gel obtained from 15% solution
Specifications
DescriptionDehydrated; Gelatin bacteriological
FormPowder
Product TypeGelatin
Quantity500 g
Unit SizeEA