Nutrient Broth (Dehydrated)
Nutrient Broth (Dehydrated)
Actual product may vary
Fisherbrand
Nutrient Broth (Dehydrated)
Cultivate fastidious pathogens and other microorganisms with this nutritious medium.
Have Questions?
Catalog NumberQuantity
FISOXCM0001B500 g
Catalog number FISOXCM0001B
Price (SGD)
-
Quantity:
500 g
Cultivate fastidious pathogens and other microorganisms with nutritious Thermo Scientific™ Oxoid™ Nutrient Broth (Dehydrated). This general purpose medium contains beef extracts combined with peptone, glucose, yeast extracts and sodium chloride to provide the nitrogen compounds, carbon, vitamins and some trace ingredients necessary for the growth of bacteria.
A general purpose fluid medium for the cultivation of microorganisms not exacting in their nutritional requirements.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand
For Laboratory Use Only
Specifications
DescriptionNutrient Broth
FormPowder
Product TypeDehydrated Culture Media
Quantity500 g
YieldFor 38.5L medium
Unit SizeEA