Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Racks
▸
Pipet Racks
▸
Hood or Wall Mount Pipette Storage Bin
Actual product may vary
Trippnt
Hood or Wall Mount Pipette Storage Bin
Have Questions?
Catalog Number
Color
FISNC9990258
White
Catalog number
FISNC9990258
Price (SGD)
-
Color:
White
Specifications
Color
White
For Use With (Equipment)
Mount with magnet or use free-standing
Product Type
Wall or Hood Pipet Bin