Zerostat 3 Anti Static Device
Zerostat 3 Anti Static Device
Actual product may vary
Zerostat 3 Anti Static Device
This Zerostat anti static gun is a pistol-shaped molded plastic device with a trigger mechanism. It does not require batteries or an electrical power supply.
Have Questions?
Catalog number FISNC9663078
Price (SGD)
-
This Zerostat anti static gun is a pistol-shaped molded plastic device with a trigger mechanism. It does not require batteries or an electrical power supply.
  • Effective in neutralizing electrostatic charges on virtually any surface
  • Eliminates static electricity in the laboratory, plant, darkroom or home
  • Prevents charged particles from flying during weighting or transfer of powders
  • Keeps powders from sticking to containers
  • Keeps film, glass, laboratory slides and plasticware dust free
  • Neutralizes static from photographic lenses, negatives and slides to release dust and lint
  • Constructed from durable hard plastic, long-lasting alternative to cleaning brushes
  • Non-radioactive
  • Convenient, hand-held and lightweight
  • Incorporates a unique piezo crystal device
  • Squeeze of the trigger creates a stream of positive ions and a stream of negative ions on trigger release, cancelling static charges
Specifications
Product TypeAnti-static Gun
For Use With (Application)Neutralizes static from photographic lenses, negatives and slides to release dust and lint
Unit SizeEA