Check Order Status
Cryomold Tissue-Tek 25 x 20 x 5 mm 100 pcs
Fisherbrand
Cryomold Tissue-Tek 25 x 20 x 5 mm 100 pcs
Cryomold Tissue-Tek 25 x 20 x 5 mm 100 pcs
Have Questions?
Catalog Number
Length (Metric)
FISNC9511236
25 mm
Catalog number
FISNC9511236
Price (SGD)
-
Length (Metric):
25 mm
Cryomold Tissue-Tek 25 x 20 x 5 mm 100 pcs
Specifications
Length (Metric)
25 mm
Width (Metric)
20 mm
Depth (Metric)
5 mm