Check Order Status
Priced Right Smiley Face Dual Dispensing Paper Towel Holder
Actual product may vary
Fisherbrand
Priced Right Smiley Face Dual Dispensing Paper Towel Holder
Have Questions?
Catalog Number
Mounting
FISNC0363518
Keyhole Slots with Double-sided Tape
Catalog number
FISNC0363518
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Mounting:
Keyhole Slots with Double-sided Tape
Specifications
Product Type
Dual Dispensing Paper Towel Holder
Color
White
Depth (English)
4 in.
Width (English)
11 in.
Height (English)
6 in.
Material
PVC
Mounting
Keyhole Slots with Double-sided Tape
For Use With (Application)
Holds one stack or about 150 multi-fold paper towels