Check Order Status
Kimwipe Holder with Magnet Mount
Actual product may vary
Kimwipe Holder with Magnet Mount
Have Questions?
Catalog Number
Mounting
FISNC0083426
Magnetic Mount
Catalog number
FISNC0083426
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Mounting:
Magnetic Mount
Specifications
Color
White
Mounting
Magnetic Mount
For Use With (Application)
Fits Kimberly Clark XL Delicate Lab Wiper Box 34155-S, or Shur-wipe 29712
Includes
Magnet Mount