Trace Elements
Actual product may vary
Trace Elements
For Research applications
Have Questions?
Catalog NumberDescription
FISMT99175CITrace Elements B
Catalog number FISMT99175CI
Price (SGD)
-
Description:
Trace Elements B
cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
  • Trace Elements A includes Cupric Sulfate, Ferric Citrate, Sodium Selenite, and Zinc Sulfate
  • Trace Elements B includes Ammonium Molybdate, Ammonium Vanadate, Manganese Sulfate, Nickel Sulfate, Sodium Silicate, Stannous Chloride and Hydrochloric Acid
  • Trace Elements C includes Aluminum Chloride, Barium Acetate, Cadmium Chloride, Chromic Chloride, Cobalt Dichloride, Germanium Dioxide, Potassium Bromide, Potassium Iodide, Rubidium Chloride, Silver Nitrate, Sodium Fluoride, and Zirconyl Chloride
Specifications
DescriptionTrace Elements B
FormLiquid
Product TypeTrace Element
SterilitySterile-filtered
FormatPlastic Bottle
Quantity100 mL
Unit SizeEach
Contents & Storage
2° to 8°C