Trypan Blue
Trypan Blue
Actual product may vary
Trypan Blue
Can be used for variety cell counting methods requiring information on cell count and viability.
Have Questions?
Catalog NumberQuantity
FISMT25900CI100 mL
Catalog number FISMT25900CI
Price (SGD)
0.00
EA
Add to cart
Quantity:
100 mL
Price (SGD)
0.00
EA
Add to cart
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
  • 0.4% (w/v) solution
  • In PBS
  • pH 7.5 ±0.5
  • Quantity: 100mL
  • Viable cells exclude the dye, while nonviable cells absorb the dye and appear blue
  • Cells should be in suspension as single cells in buffered saline before counting
  • Can be used in assays providing total cell count, viable cell or nonviable cell count alone
  • Solution is provided at the common concentration of 0.4% (w/v)
Specifications
Solution TypeTrypan Blue 0.4% Solution in PBS
Quantity100 mL
FormatLiquid
Storage Requirements15° to 30°C
Packaging TypePlastic Bottle
pH7.5 ±0.5
Unit SizeEA