Lymphocyte Separation Medium
Lymphocyte Separation Medium
Actual product may vary
Lymphocyte Separation Medium
Designed for the in vitro isolation of lymphocytes from diluted whole blood.
Have Questions?
Catalog NumberQuantity
FISMT25072CV1 x 500 mL
Catalog number FISMT25072CV
Price (SGD)
-
Quantity:
1 x 500 mL
Lymphocyte Separation Medium was originally designed for the in vitro isolation of lymphocytes from diluted whole blood. It is a sterile filtered, iso-osmotic polysucrose and diatrizoate solution with low viscosity and a density of 1.077-1.080 g/cm3 at 20°C.
Specifications
FormLiquid
Product TypeLymphocyte separation medium
SterilitySterile
Quantity1 x 500 mL
Unit SizeEA
Contents & Storage
2° to 30°C