Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
Cell Culture Grade Water Tested to USP Sterile Water for Injection Specifications, Corning™
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP Sterile Water are used in preparation of purification and chromatography buffers.
Have Questions?
Change viewbuttonViewtableView
Quantity:
6 x 100 mL
6 x 500 mL
Packaging:
Bottle
Catalog number FISMT25055CI
Price (SGD)
-
Quantity:
6 x 100 mL
Packaging:
Bottle
Request bulk or custom format
Specifications
PackagingBottle
Quantity6 x 100 mL
Unit SizePK
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).