Cell Culture Phosphate Buffered Saline (1X)
Cell Culture Phosphate Buffered Saline (1X)
Actual product may vary
Cell Culture Phosphate Buffered Saline (1X)
With low endotoxin content. Without calcium and magnesium.
Have Questions?
Catalog NumberQuantity
FISMT21040CV6 x 500 mL
Catalog number FISMT21040CV
Price (SGD)
558.00
PK
Add to cart
Quantity:
6 x 500 mL
Price (SGD)
558.00
PK
Add to cart
Phosphate buffered saline (PBS) is a buffer solution containing sodium chloride, sodium phosphate, and in some formulations potassium chloride and potassium phosphate. PBS helps to maintain a constant pH and is isotonic and non-toxic to cells. This formulation does not contain calcium and magnesium.
Specifications
Dilution1 X
Quantity6 x 500 mL
FormulationPotassium Dihydrogen Phosphate 0.144g/L, Sodium Chloride 9g/L, Disodium Phosphate 0.795g/L
pH7.3 to 7.5
FormLiquid
Unit SizePK
Contents & Storage
2° to 30°C