Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
Actual product may vary
Cell Culture Buffers: Dulbecco's Phosphate-Buffered Salt Solution 1X
With low endotoxin content.
Have Questions?
Catalog NumberQuantityProduct TypeFormAdditive
FISMT21031CV6 x 500 mLDPBSLiquidNo Calcium, No Magnesium
Catalog number FISMT21031CV
Price (SGD)
-
Quantity:
6 x 500 mL
Product Type:
DPBS
Form:
Liquid
Additive:
No Calcium, No Magnesium
Dulbecco's Phosphate Buffered Saline (DPBS) is a buffer solution which can be used to maintain cell culture media in the physiological pH range of 7.0 to 7.6. It can serve as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time. 
Specifications
AdditiveNo Calcium, No Magnesium
Dilution1 X
Product TypeDPBS
Quantity6 x 500 mL
FormulationPotassium Chloride 0.2g/L, Potassium Dihydrogen Phosphate 0.2g/L, Sodium Chloride 8g/L, Disodium Phosphate 1.15g/L
pH7.3 to 7.5
FormLiquid
Unit SizePK
Contents & Storage
2° to 30°C