Hank's Balanced Salt Solutions
Hank's Balanced Salt Solutions
Actual product may vary
Hank's Balanced Salt Solutions
Salt solutions are shipped at ambient temperature unless otherwise indicated
Have Questions?
Catalog NumberQuantityConcentrationWith AdditivesWithout Additives
FISMT21023CM6 x 1 L1 XCalcium, MagnesiumNo Phenol Red
Catalog number FISMT21023CM
Price (SGD)
721.00
PK
Add to cart
Quantity:
6 x 1 L
Concentration:
1 X
With Additives:
Calcium, Magnesium
Without Additives:
No Phenol Red
Price (SGD)
721.00
PK
Add to cart
pH of 10X solutions must be adjusted following dilution to 1X concentration.
Specifications
Concentration1 X
FormLiquid
Product TypeHanks' Balanced Salt Solution
Shelf Life3 Years
With AdditivesCalcium, Magnesium
Without AdditivesNo Phenol Red
FormulationCalcium Chloride (anhydrous) 0.14g/L, Potassium Chloride 0.4g/L , Monopotassium Phosphate 0.06g/L, Magnesium Sulfate (anhydrous) 0.977g/L, Sodium Chloride 8g/L , Disodium Phosphate (anhydrous) 0.0477g/L, Sodium Bicarbonate 0.35g/L, D-Glucose 1g/L
pH7.1 to 7.4
Quantity6 x 1 L
Unit SizePK
Contents & Storage
2° to 30°C