RPMI 1640 Medium (Mod.) 1X with L-Glutamine
RPMI 1640 Medium (Mod.) 1X with L-Glutamine
Actual product may vary
RPMI 1640 Medium (Mod.) 1X with L-Glutamine
Guaranteed low endotoxin levels ≤0.025EU/mL
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityWith AdditivesWithout Additives
FISMT10040CM6 × 1 LGlutamineNone
CRG10-040-CM6 × 1 LGlutamineNone
Catalog number FISMT10040CM
Price (SGD)
-
Quantity:
6 × 1 L
With Additives:
Glutamine
Without Additives:
None
  • Ready-to-use media for mammalian in vitro cell culture is supplied in polyethylene teraphthalate (PET) plastic bottles
  • Sonically welded cap liners eliminate toxic resins or glues that could leach into media
  • Shipped at ambient temperature
Specifications
FormLiquid
Product TypeRPMI 1640
With AdditivesGlutamine
Without AdditivesNone
Quantity6 × 1 L
pH7.2 to 7.4
Unit SizePack
Contents & Storage
2° to 8°C