Check Order Status
Microbial Count Samplers
Actual product may vary
MilliporeSigma
Microbial Count Samplers
Sterilized, individually packaged, ready-to-use dip testers for Total Count and HPC Total Count
Have Questions?
Catalog Number
Pore Size
FISMHPC10025
0.45μm
Catalog number
FISMHPC10025
Price (SGD)
807.00
PK
Add to cart
Pore Size:
0.45μm
Price (SGD)
807.00
PK
Add to cart
Sterilized, individually packaged, ready-to-use dip testers for Total Count and HPC Total Count
Convenient alternative to Standard Methods MF method for routine monitoring of microbiological organisms
Consist of plastic paddle with air vent, filter in contact with nutrient-containing pad, and plastic sampling/incubation case
0.45μ filter is marked with a grid to aid in counting colonies on its surface
Incubation time: 48 to 72 hours at 25° to 35°C (77° to 95°F)
Magnification of colonies is recommended.
Specifications
Color
Red
Filter Material
Mixed Cellulose Esters
For Use With (Application)
Clinical lab, environmental monitoring, food and beverage, water monitoring
Incubation Temperature
25°C to 35°C
Material
SAN
Pore Size
0.45μm
Storage Requirements
15°C to 30°C