Check Order Status
Buffer Solution pH 7.00 ±0.01 Yellow, MilliporeSigma™
Element Search
Structure Search
Buffer Solution pH 7.00 ±0.01 Yellow, MilliporeSigma™
Have Questions?
Catalog Number
Quantity
FISMBX16321
500 mL
Catalog number
FISMBX16321
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Material
Buffer Solution pH 7.00 ±0.01
Name Note
Yellow
CAS
7558-79-4
Color
Yellow
Recommended Storage
+15°C to +3°C
View more