Check Order Status
Malachite Green Oxalate (Certified Biological Stain), Fisher Chemical
Element Search
Structure Search
Malachite Green Oxalate (Certified Biological Stain), Fisher Chemical
Have Questions?
Catalog Number
Quantity
FISM290-25
25 g
Catalog number
FISM290-25
Price (SGD)
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Material
Malachite Green Oxalate
Melting Point
164°C
CAS
2437-29-8
ChemAlert Storage Symbol
Blue
Bio. Stain Comm. Certification
Pass Test
View more