MQuant™ Chlorine Test Strips
MQuant™ Chlorine Test Strips
Actual product may vary
MilliporeSigma
MQuant™ Chlorine Test Strips
To determine the presence of residues of chlorine
Have Questions?
Catalog NumberRangeNo. of Tests
FISM11792500010.5 to 20mg/L75
Catalog number FISM1179250001
Price (SGD)
-
Range:
0.5 to 20mg/L
No. of Tests:
75
  • Small and easy to handle
  • Flexible and universal application
  • Redox reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable
  • Color-scale graduation: 0.5 - 1 - 2 - 5 - 10 - 20mg/L
Reclose the tube containing the test strips immediately after use
For water analysis
Specifications
For Use With (Application)To check for presence of chlorine in disinfectants and water
For Use With (Equipment)Colorimeter
Graduations0.5-1-2-5-10-20mg/L
No. of Tests75
Range0.5 to 20mg/L
Storage Requirements2°C to 8°C
Test MethodRedox Reaction
TypeTest Strip
Unit SizeEA