MQuant™ Peroxide Test Strips
Actual product may vary
MilliporeSigma
MQuant™ Peroxide Test Strips
To check for presence or absence of residues of Peroxide
Have Questions?
Catalog NumberQuantityRange
FISM1100110001100 Strips/Pk.0.5 to 25 mg/L H2O2
Catalog number FISM1100110001
Price (SGD)
230.00
Each
Quantity:
100 Strips/Pk.
Range:
0.5 to 25 mg/L H2O2
Price (SGD)
230.00
Each
  • Small and easy to handle
  • Flexible and universal application
  • Enzymatic reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable
1.10011.0001
Color-scale graduation: 0.5 - 2 - 5 - 10 - 25mg/L H2O2
1.10081.0001
Color-scale graduation: 1 - 3 - 10 - 30 - 100mg/L H2O2
1.10337.0001
Color-scale graduation: 100 - 200 - 400 - 600 - 800 - 1000mg/L H2O2
Reclose the tube containing the test strips immediately after use.
For food and water analysis
Specifications
For Use With (Application)For testing residual concentrations of disinfectants on equipment or the product itself
For Use With (Equipment)Colorimeter
Graduations0.5-2-5-10-25
No. of Tests100
Quantity100 Strips/Pk.
Range0.5 to 25 mg/L H2O2
Storage Requirements2°C to 8°C
Test MethodEnzymatic Reaction
TypePeroxide Test Strips
Unit SizeEach